Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenoxyacetic acid hydrazide, 98%

$72.32 - $289.30

Chemical Identifiers

Chemical Name or Material 2-Chlorophenoxyacetic acid hydrazide
CAS 36304-40-2
Molecular Formula C8H9ClN2O2
Formula Weight 200.63
MDL Number MFCD00082786
Beilstein 979183
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1358206

  SDS
alfa aesar™
A1358206
5g Each for $72.32

AAA1358214

  SDS
alfa aesar™
A1358214
25g Each for $289.30
Description & Specifications

Specifications

Formula Weight 200.63
Chemical Name or Material 2-Chlorophenoxyacetic acid hydrazide
MDL Number MFCD00082786
Beilstein 979183
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Health Hazard 1 Warning
CAS 36304-40-2
Molecular Formula C8H9ClN2O2
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Melting Point 110° to 113°C
Quantity 5g
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.