Please login to your online account to display your discounted pricing

2-Chlorophenothiazine, 99%, ACROS Organics™

$42.36 - $42.36

Chemical Identifiers

Chemical Name or Material 2-Chlorophenothiazine
Name Note 99%
CAS 92-39-7
CAS Min % 98.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 99%
Molecular Formula C12H8ClNS
Formula Weight 233.71
MDL Number MFCD00005016
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC334550250

  SDS
thermo fisher scientific
334550250
Glass bottle 25g Each for $42.36
Description & Specifications

Specifications

Assay ≥98.5%
Physical Form Crystalline Powder
Formula Weight 233.71
Quantity 25g
Color Pale green to yellow
TSCA TSCA
EINECS Number 202-152-5
RTECS Number SN7580000
Melting Point 200° to 206°C
CAS Max % 100.0
CAS Min % 98.5
CAS 92-39-7
Molecular Formula C12H8ClNS
MDL Number MFCD00005016
Name Note 99%
Chemical Name or Material 2-Chlorophenothiazine
Packaging Glass bottle
Assay Percent Range 99%
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.