Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorophenothiazine, 99%

$49.42 - $145.70

Chemical Identifiers

Chemical Name or Material 2-Chlorophenothiazine
CAS 92-39-7
Molecular Formula C12H8ClNS
Formula Weight 233.72
MDL Number MFCD00005016
Beilstein 176587
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2143814

  SDS
alfa aesar™
B2143814
25g Each for $49.42

AAB2143822

  SDS
alfa aesar™
B2143822
100g Each for $145.70
Description & Specifications

Specifications

Formula Weight 233.72
Melting Point 197° to 202°C
TSCA TSCA
Health Hazard 1 Warning
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
CAS 92-39-7
Beilstein 176587
MDL Number MFCD00005016
Quantity 25g
Chemical Name or Material 2-Chlorophenothiazine
Odor Odorless
Molecular Formula C12H8ClNS
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
RTECS Number SN7580000
Assay 99%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.