missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorophenol, 98+%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals 180995000

 View more versions of this product

Catalog No. AC180995000



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-Chlorophenol
95-57-8
100.0
1.2620g/mL
63°C
Glass bottle
1.5580 to 1.5600
MFCD00002159
06,183
15,2157
Solubility in water: 28.5g/l (20°C). Other solubilities: freely soluble in solutions of sodium hydroxide,, alcohol and ether, soluble in aromatic solvents
C1=CC=C(C(=C1)O)Cl
128.56
4.11 mPa.s (25°C)
128.56
98+%
98.0
8.0°C
173.0°C to 175.0°C
98% min. (GC)
C6H5ClO
ClC6H4OH
500 g
1.262
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
ISPYQTSUDJAMAB-UHFFFAOYSA-N
2-chlorophenol
7245
CHEBI:47083
98+%

Chemical Identifiers

95-57-8
128.56
ISPYQTSUDJAMAB-UHFFFAOYSA-N
7245
2-chlorophenol
C6H5ClO
MFCD00002159
o-chlorophenol, phenol, 2-chloro, 2-hydroxychlorobenzene, o-chlorphenol, o-chlorophenic acid, phenol, o-chloro, 2-chloro-1-hydroxybenzene, chlorophenol, septi-kleen, 2-chloro-phenol
CHEBI:47083
C1=CC=C(C(=C1)O)Cl
Safety and Handling

Safety and Handling

GHS H Statement
Toxic to aquatic life with long lasting effects.
Harmful in contact with skin.
Harmful if swallowed.
Harmful if inhaled.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for severa

GHS Signal Word: Warning

EINECSNumber : 202-433-2

RTECSNumber : SK2625000

TSCA : TSCA

SDS Documents

Documents

Promotions

Promotions

RUO – Research Use Only