missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorophenethylalcohol, 98%, ACROS Organics™

Chemical Identifiers

CAS 19819-95-5
Molecular Formula C8H9ClO
Molecular Weight (g/mol) 156.609
MDL Number MFCD00002888
InChI Key IWNHTCBFRSCBQK-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
PubChem CID 88266
IUPAC Name 2-(2-chlorophenyl)ethanol
SMILES C1=CC=C(C(=C1)CCO)Cl
View More Specs

Products 3
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC186190010 SDS View Documents Acros Organics
AC186190010
1g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC186190250 SDS View Documents Acros Organics
186190250
25g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AC186190050 SDS View Documents Acros Organics
186190050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

19819-95-5
156.609
IWNHTCBFRSCBQK-UHFFFAOYSA-N
88266
C1=CC=C(C(=C1)CCO)Cl
C8H9ClO
MFCD00002888
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
2-(2-chlorophenyl)ethanol

Specifications

1.1900g/mL
Authentic
1.5495 to 1.5515
84.0°C to 85.0°C (3.0mmHg)
1g
97.5
98%
ClC6H4CH2CH2OH
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol
C1=CC=C(C(=C1)CCO)Cl
156.609
156.61
98%
>110°C
Glass bottle
1.19
Colorless
19819-95-5
100.0
C8H9ClO
MFCD00002888
IWNHTCBFRSCBQK-UHFFFAOYSA-N
2-(2-chlorophenyl)ethanol
88266
Liquid
2-Chlorophenethylalcohol, 98%
Safety and Handling

Safety and Handling

EINECSNumber : 243-348-0

TSCA : TSCA