missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chlorophenethylalcohol, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 186190010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chlorophenethylalcohol | |
19819-95-5 | |
97.5 | |
1.1900g/mL | |
1 g | |
Glass bottle | |
1.5495 to 1.5515 | |
1.19 | |
ClC6H4CH2CH2OH | |
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol | |
C1=CC=C(C(=C1)CCO)Cl | |
156.61 | |
156.61 |
98% | |
Colorless | |
100.0 | |
84.0°C to 85.0°C (3.0 mmHg) | |
Authentic | |
Liquid | |
97.5% min. (GC) | |
C8H9ClO | |
MFCD00002888 | |
IWNHTCBFRSCBQK-UHFFFAOYSA-N | |
2-(2-chlorophenyl)ethanol | |
88266 | |
98% |
Chemical Identifiers
19819-95-5 | |
156.61 | |
IWNHTCBFRSCBQK-UHFFFAOYSA-N | |
88266 | |
C1=CC=C(C(=C1)CCO)Cl |
C8H9ClO | |
MFCD00002888 | |
2-2-chlorophenyl ethanol, 2-chlorophenethyl alcohol, benzeneethanol, 2-chloro, 2-chlorophenethylalcohol, 2-2-chlorophenyl ethan-1-ol, o-chlorophenethyl alcohol, o-chlorophenethylic alcohol, o-chlorophenylmethylcarbinol, chlorobenzene-ethanol, 2-chlorobenzeneethanol | |
2-(2-chlorophenyl)ethanol |
Safety and Handling
EINECSNumber : 243-348-0
TSCA : TSCA
RUO – Research Use Only