Please login to your online account to display your discounted pricing

2-Chloronicotinoyl chloride, +98%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC395790250

 View more versions of this product

Catalog No. AC395790250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Physical Form Low Melting Solid
Assay ≥98.0% (derivatized)
Formula Weight 176
Quantity 25g
Color White to beige-yellow
Melting Point 39° to 44°C
CAS Max % 100.0
CAS Min % 96.0
Assay Percent Range 97%
Molecular Formula C6H3Cl2NO
Synonym 2-Chloropyridine-3-carbonyl chloride
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-Chloronicotinoyl chloride
MDL Number MFCD00051677
CAS 49609-84-9
NMR Consistent with structure
Packaging Glass bottle

Specifications

Chemical Name or Material 2-Chloronicotinoyl chloride
CAS 49609-84-9
CAS Min % 96.0
CAS Max % 100.0
Assay ≥98.0% (derivatized)
Assay Percent Range 97%
Molecular Formula C6H3Cl2NO
Formula Weight 176
MDL Number MFCD00051677
Synonym 2-Chloropyridine-3-carbonyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Reacts violently with water.

SDS & Safety Documents