missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloronicotinamide, 98%, ACROS Organics™

$317.35 - $317.35

Chemical Identifiers

CAS 10366-35-5
Molecular Formula C6H5ClN2O
Molecular Weight (g/mol) 156.569
MDL Number MFCD00006237
InChI Key ZQZAHPFFZWEUCL-UHFFFAOYSA-N
Synonym 2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinamide, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
PubChem CID 82588
IUPAC Name 2-chloropyridine-3-carboxamide
SMILES C1=CC(=C(N=C1)Cl)C(=O)N
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC185760250 SDS View Documents Acros Organics
185760250
25g Glass bottle Each for $317.35
AC185760050 SDS View Documents Acros Organics
185760050
5g Glass bottle N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Chemical Identifiers

10366-35-5
156.569
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
82588
C1=CC(=C(N=C1)Cl)C(=O)N
C6H5ClN2O
MFCD00006237
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinamide, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
2-chloropyridine-3-carboxamide

Specifications

Authentic
Beige
25g
97.5
98%
Cl(C5H3N)CONH2
22, II, 36
ZQZAHPFFZWEUCL-UHFFFAOYSA-N
2-chloropyridine-3-carboxamide
82588
Fine Crystalline Powder
2-Chloronicotinamide, 98%
Glass bottle
162°C to 166°C
10366-35-5
100.0
C6H5ClN2O
MFCD00006237
2-chloronicotinamide, 3-pyridinecarboxamide, 2-chloro, chloronicotinamide, 2-chloro-nicotinamide, acmc-1bvtu, nicotinamide, 2-chloro, 2-chloronicotinamide, 2-chloronicotinic acid amide, ksc492e7f, 2-chloro-3-pyridinecarboxamide
C1=CC(=C(N=C1)Cl)C(=O)N
156.569
156.57
≥97.5%
Safety and Handling

Safety and Handling

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Causes skin irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 233-808-9

TSCA : TSCA