Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloronicotinamide, 98+%

$26.80 - $230.89

Chemical Identifiers

Chemical Name or Material 2-Chloronicotinamide
CAS 10366-35-5
Molecular Formula C6H5ClN2O
Formula Weight 156.57
MDL Number MFCD00006237
Beilstein 119026
Synonym 2-Chloropyridine-3-carboxamide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1209003

 
alfa aesar™
A1209003
1g Each for $26.80

AAA1209006

 
alfa aesar™
A1209006
5g Each for $71.27

AAA1209014

 
alfa aesar™
A1209014
25g Each for $230.89
Description & Specifications

Specifications

Formula Weight 156.57
Chemical Name or Material 2-Chloronicotinamide
Melting Point 163° to 167°C
Quantity 1g
Assay ≥98%
Molecular Formula C6H5ClN2O
MDL Number MFCD00006237
CAS 10366-35-5
Beilstein 119026
Synonym 2-Chloropyridine-3-carboxamide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.