Please login to your online account to display your discounted pricing

2-Chloromethylbenzimidazole 95%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloromethylbenzimidazole
Name Note 95%
CAS 1080123
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C8H7ClN2
Formula Weight 166.61
MDL Number MFCD00005602
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AC109390050

 
fisher scientific acros
109390050
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC109390250

 
fisher scientific acros
109390250
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 166.61
Physical Form Powder
Assay ≥94%
Quantity 5g
Color Beige to dark yellow
Melting Point 145° to 150°C
EINECS Number 225-452-8
TSCA TSCA
CAS 1080123
CAS Min % 94.0
CAS Max % 100.0
Chemical Name or Material 2-Chloromethylbenzimidazole
Infrared Spectrum Authentic
Health Hazard 3 GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Name Note 95%
Assay Percent Range 95%
Molecular Formula C8H7ClN2
MDL Number MFCD00005602
Health Hazard 2 GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.
Packaging Glass bottle
Safety and Handling

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
Harmful if swallowed.