Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Chloromethyl)tetrahydropyran, 98%

Chemical Identifiers

Chemical Name or Material 2-(Chloromethyl)tetrahydropyran
CAS 18420-41-2
Molecular Formula C6H11ClO
Formula Weight 134.61
MDL Number MFCD00006622
UN Number UN1993
Beilstein 102996
Synonym (2-Tetrahydropyranyl)methyl chloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0772506

  SDS
alfa aesar™
L0772506
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAL0772514

  SDS
alfa aesar™
L0772514
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 134.61
Chemical Name or Material 2-(Chloromethyl)tetrahydropyran
Assay 98%
Molecular Formula C6H11ClO
MDL Number MFCD00006622
CAS 18420-41-2
UN Number UN1993
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
Boiling Point 53° to 54°C (12mmHg)
Beilstein 102996
Synonym (2-Tetrahydropyranyl)methyl chloride
Flash Point 55°C (131°F)
Quantity 5g

Applications
2-(Chloromethyl)tetrahydropyran can be used to produce hex-5-en-1-ol. This reaction will need reagents of sodium, diethyl ether.

Solubility
Difficult to mix.

Notes
Store away from strong oxidizing agents. Keep container tightly closed. Store in cool, dry conditions in well sealed containers.

Safety and Handling

Warning

GHS P Statement
P210-P280-P240-P303+P361+P353-P403+P235-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
Ground/bond container and receiving equipment.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
Store in a well-ventilated place. Keep cool.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H226
Flammable liquid and vapor.