missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Chloromethyl)quinoline Hydrochloride 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific 109510250

 View more versions of this product

Catalog No. AC109510250

 

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications Description & Specifications

Specifications

2-(Chloromethyl)quinoline hydrochloride
97%
96.0
Authentic
Powder
Pale yellow to cream or light pink
25g
214.1
(5% aq. soln.) clear
3747-74-8
100.0
Glass bottle
≥96.0%
97%
C10H8ClN·HCl
MFCD00012734

Chemical Identifiers

3747-74-8
100.0
C10H8ClN·HCl
MFCD00012734
≥96.0%
96.0
97%
214.1
2-(Chloromethyl)quinoline hydrochloride, 97%

Safety and Handling

SDS & Safety Documents

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

EINECSNumber : 223-145-3

TSCA : TSCA