missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Chloromethyl)pyridine hydrochloride, 98%

Manufacturer:  Alfa Aesar™ A1022622

 View more versions of this product

Catalog No. AAA1022622



Description

Description

Applications
2-(Chloromethyl)pyridine hydrochloride was used as reagent in base catalyzed alkylation of p-tert-butylcalix[6]arene and p-tert-butylcalix[5]arene in DMF. It was used in the synthesis of Gd3+ diethylenetriaminepentaacetic acid bisamide complex, a Zn2+-sensitive magnetic resonance imaging contrast agent.

Solubility
Soluble in water, acetone (10-50mg/ml), 95%ethanol (100mg/ml).

Notes
Hygroscopic store in inert gas Store away from strong oxidizing agents. keep container tightly closed. Store in cool, dry conditions in well sealed containers.
Specifications

Specifications

2- (Chloromethyl)pyridine hydrochloride
100g
C6H7Cl2N
UN3261
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-chloromethyl pyridine hydrochloride, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
C1=CC=NC(=C1)CCl.Cl
164.029
CHEBI:76600
98%
120°C to 124°C
6959-47-3
MFCD00012811
3689155
JPMRGPPMXHGKRO-UHFFFAOYSA-N
2-(chloromethyl)pyridine;hydrochloride
23392
164.04

Chemical Identifiers

6959-47-3
164.029
JPMRGPPMXHGKRO-UHFFFAOYSA-N
23392
2-(chloromethyl)pyridine;hydrochloride
C6H7Cl2N
MFCD00012811
2-chloromethyl pyridine hydrochloride, 2-picolyl chloride hydrochloride, 2-chloromethylpyridine hydrochloride, picolyl chloride hydrochloride, 2-chloromethyl pyridine hcl, 2-chloromethyl pyridine hydrochloride, 2-pyridylmethylchloride hydrochloride, 2-chloromethyl pyridinium chloride, pyridine, 2-chloromethyl-, hydrochloride, unii-yni4417kx0
CHEBI:76600
C1=CC=NC(=C1)CCl.Cl
Safety and Handling

Safety and Handling

GHS H Statement
H301-H311-H314-H318-H317
Toxic if swallowed.
Toxic in contact with skin.
Causes severe skin burns and eye damage.
Causes serious eye damage.
May cause an allergic skin reaction.

GHS P Statement
P260-P301+P310-P303+P361+P353-P305+P351+P338-P361-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
Continue rinsing.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : US6825000

SDS Documents