Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Chloromethyl)phenyl isocyanate, 97%

Chemical Identifiers

Chemical Name or Material 2-(Chloromethyl)phenyl isocyanate
CAS 52986-66-0
Molecular Formula C8H6ClNO
Formula Weight 167.6
MDL Number MFCD00077427
UN Number UN2922
Beilstein 957161
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1599503

  SDS
alfa aesar™
L1599503
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 167.6
Chemical Name or Material 2-(Chloromethyl)phenyl isocyanate
Molecular Formula C8H6ClNO
CAS 52986-66-0
Health Hazard 2 GHS H Statement
H331-H334-H314-H318-H302-H312-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.
MDL Number MFCD00077427
UN Number UN2922
Beilstein 957161
Health Hazard 1 Danger
Health Hazard 3 GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUIDS, TOXIC, N.O.S.
Flash Point >110°C (230°F)
Boiling Point 43° to 45°C (0.2mmHg)
Quantity 1g
Assay 97%
Recommended Storage 2° to 8°C
Safety and Handling

Danger

GHS P Statement
P260-P284a-P303+P361+P353-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H331-H334-H314-H318-H302-H312-H317
Toxic if inhaled.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.
Harmful if swallowed.
Harmful in contact with skin.
May cause an allergic skin reaction.

2° to 8°C