Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-(Chloromethyl)benzimidazole, 96%

$52.34 - $195.49

Chemical Identifiers

Chemical Name or Material 2-(Chloromethyl)benzimidazole
CAS 4857-04-9
Molecular Formula C8H7ClN2
Formula Weight 166.61
MDL Number MFCD00005602
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2554806

  SDS
alfa aesar™
B2554806
5g Each for $52.34

AAB2554814

  SDS
alfa aesar™
B2554814
25g Each for $195.49
Description & Specifications

Specifications

Formula Weight 166.61
Chemical Name or Material 2-(Chloromethyl)benzimidazole
Assay 96%
Molecular Formula C8H7ClN2
MDL Number MFCD00005602
CAS 4857-04-9
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 141° to 143°C (dec.)
Quantity 5g

Applications
2-(Chloromethyl)benzimidazole is used as intermediate in pharmaceutical , in organic synthesis .

Solubility
Insoluble in water.

Notes
Store in cool, dry place in a well sealed container. Store away from oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.