missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Chloromethyl)-5-phenyl-1,3,4-oxadiazole, 97%, ACROS Organics™

Manufacturer:  Thermo Fisher Scientific AC438280050

 View more versions of this product

Catalog No. AC438280050

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Specifications

Specifications

2-(chloromethyl)-5-phenyl-1, 3, 4-oxadiazole
White
5g
96.0
96% min. (GC)
1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, chloromethylphenyloxadiazole, enamine_005035
C1=CC=C(C=C1)C2=NN=C(O2)CCl
194.62
194.62
97%
Authentic
119.3°C to 119.7°C
33575-83-6
100.0
C9H7ClN2O
AGLNTFQAHIRTFA-UHFFFAOYSA-N
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
314941
Crystalline Powder

Chemical Identifiers

33575-83-6
194.62
1,3,4-oxadiazole, 2-chloromethyl-5-phenyl, 1,3,4-oxadiazole,2-chloromethyl-5-phenyl, 2-chloromethyl-5-phenyl-1,3,4 oxadiazole, 2-chloromethyl-5-phenyl-1,3,4-oxadiazole, 5-chloromethyl-1,3,4-oxadiazol-2-yl benzene, 5-chloromethyl-2-phenyl-1,3,4-oxadiazole, chloromethylphenyloxadiazole, enamine_005035
2-(chloromethyl)-5-phenyl-1,3,4-oxadiazole
C9H7ClN2O
AGLNTFQAHIRTFA-UHFFFAOYSA-N
314941
C1=CC=C(C=C1)C2=NN=C(O2)CCl
Safety and Handling

Safety and Handling

GHS H Statement
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: If breathing is difficult,remove to fresh air and keep at rest in a position comfortable for breathing.

GHS Signal Word: Warning

SDS Documents