Please login to your online account to display your discounted pricing

2-(Chloromethyl)-5-(4-methylphenyl)-1,3,4-oxadiazole, ≥95%, Maybridge


Quantity 5g

Manufacturer: fisher scientific company  RF02900DE

 View more versions of this product

Catalog No. RF02900DE

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 208.65
Quantity 5g
Assay 95%
CAS Min % 95.0
Molecular Formula C10H9ClN2O
Assay Percent Range 95%
MDL Number MFCD00662318
CAS 287197-95-9
Chemical Name or Material 2-(Chloromethyl)-5-(4-methylphenyl)-1, 3, 4-oxadiazole
Health Hazard 2 GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Chloromethyl)-5-(4-methylphenyl)-1, 3, 4-oxadiazole
CAS 287197-95-9
CAS Min % 95.0
Assay 95%
Assay Percent Range 95%
Molecular Formula C10H9ClN2O
Formula Weight 208.65
MDL Number MFCD00662318
Accessories

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.