Please login to your online account to display your discounted pricing

2-(Chloromethyl)-2-methyloxirane, 97%, Maybridge


Quantity 10g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CD08593EA

 View more versions of this product

Catalog No. CD08593EA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 106.55
Quantity 10g
Assay 97%
CAS Max % 100.0
Health Hazard 1 Danger
Chemical Name or Material 2-(Chloromethyl)-2-methyloxirane
Assay Percent Range 97%
MDL Number MFCD00052487
CAS 598-09-4
CAS Min % 96.5
Molecular Formula C4H7ClO
Packaging Amber Glass Bottle
Health Hazard 2 GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-(Chloromethyl)-2-methyloxirane
CAS 598-09-4
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C4H7ClO
Formula Weight 106.55
MDL Number MFCD00052487
Accessories

Danger

GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS P Statement
Causes severe skin burns and eye damage.
Harmful if swallowed.
Harmful in contact with skin.
Causes serious eye damage.
Harmful if inhaled.

SDS & Safety Documents