Please login to your online account to display your discounted pricing

2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride, 97%, Maybridge


Quantity 1g
Packaging Amber Glass Bottle

Manufacturer: fisher scientific company  CC20874DA

 View more versions of this product

Catalog No. CC20874DA

  • / Each
 

Description & Specifications

Chemical Identifiers

Formula Weight 167.04
Quantity 1g
Assay 97%
CAS 78667-04-6
Assay Percent Range 97%
Molecular Formula C5H7ClN2·ClH
Packaging Amber Glass Bottle
Chemical Name or Material 2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-(Chloromethyl)-1-methyl-1H-imidazole hydrochloride
Assay 97%
Assay Percent Range 97%
Molecular Formula C5H7ClN2·ClH
Formula Weight 167.04
Accessories

GHS Signal Word: Danger

SDS & Safety Documents