Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloromethyl-1,4-benzodioxane, 97%

$116.99 - $469.49

Chemical Identifiers

Chemical Name or Material 2-Chloromethyl-1,4-benzodioxane
CAS 2164-33-2
Molecular Formula C9H9ClO2
Formula Weight 184.62
MDL Number MFCD00014666
Beilstein 150153
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0413706

  SDS
alfa aesar™
L0413706
5g Each for $116.99

AAL0413714

  SDS
alfa aesar™
L0413714
25g Each for $469.49
Description & Specifications

Specifications

Formula Weight 184.62
Chemical Name or Material 2-Chloromethyl-1,4-benzodioxane
MDL Number MFCD00014666
Assay 97%
Molecular Formula C9H9ClO2
Beilstein 150153
Boiling Point 96°C (0.4mmHg)
Quantity 5g
CAS 2164-33-2