missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloromandelic acid, 98%, ACROS Organics™

$215.77 - $215.77

Chemical Identifiers

CAS 10421-85-9
Molecular Formula C8H7ClO3
Molecular Weight (g/mol) 186.59
MDL Number MFCD00084962
InChI Key RWOLDZZTBNYTMS-UHFFFAOYSA-N
Synonym 2-2-chlorophenyl-2-hydroxy-acetic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, 2-chloro-mandelic acid, 2-chloromandelic acid, 2-chloromandelicacid, 2-chlorophenyl glycolic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, dl-2-chloromandelic acid, pubchem15895
PubChem CID 97720
IUPAC Name 2-(2-chlorophenyl)-2-hydroxyacetic acid
SMILES C1=CC=C(C(=C1)C(C(=O)O)O)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC240680500
SDS View Documents
Thermo Fisher Scientific
240680500
50g Glass bottle
Each for $215.77
 
AC240682500
SDS View Documents
Thermo Fisher Scientific
AC240682500
250g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

10421-85-9
186.59
RWOLDZZTBNYTMS-UHFFFAOYSA-N
97720
C1=CC=C(C(=C1)C(C(=O)O)O)Cl
C8H7ClO3
MFCD00084962
2-2-chlorophenyl-2-hydroxy-acetic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, 2-chloro-mandelic acid, 2-chloromandelic acid, 2-chloromandelicacid, 2-chlorophenyl glycolic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, dl-2-chloromandelic acid, pubchem15895
2-(2-chlorophenyl)-2-hydroxyacetic acid

Specifications

Authentic
Cream to White
50g
97.5
97.5% min. (HPLC)
ClC6H4CH(OH)COOH
2-2-chlorophenyl-2-hydroxy-acetic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, 2-chloro-mandelic acid, 2-chloromandelic acid, 2-chloromandelicacid, 2-chlorophenyl glycolic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, dl-2-chloromandelic acid, pubchem15895
C1=CC=C(C(=C1)C(C(=O)O)O)Cl
186.59
186.59
98%
Glass bottle
80°C to 85°C
10421-85-9
100.0
C8H7ClO3
MFCD00084962
RWOLDZZTBNYTMS-UHFFFAOYSA-N
2-(2-chlorophenyl)-2-hydroxyacetic acid
97720
Crystalline Powder
2-Chloromandelic acid
Safety and Handling

Safety and Handling

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 233-900-9

SDS