Learn More
2-chloromandelic acid, 98%, ACROS Organics™
Manufacturer: Thermo Fisher Scientific 240680500
Specifications
2-Chloromandelic acid | |
Glass bottle | |
80°C to 85°C | |
10421-85-9 | |
100.0 | |
C8H7ClO3 | |
MFCD00084962 | |
RWOLDZZTBNYTMS-UHFFFAOYSA-N | |
2-(2-chlorophenyl)-2-hydroxyacetic acid | |
97720 | |
Crystalline Powder |
Authentic | |
Cream to White | |
50g | |
97.5 | |
97.5% min. (HPLC) | |
ClC6H4CH(OH)COOH | |
2-2-chlorophenyl-2-hydroxy-acetic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, 2-chloro-mandelic acid, 2-chloromandelic acid, 2-chloromandelicacid, 2-chlorophenyl glycolic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, dl-2-chloromandelic acid, pubchem15895 | |
C1=CC=C(C(=C1)C(C(=O)O)O)Cl | |
186.59 | |
186.59 | |
98% |
Chemical Identifiers
10421-85-9 | |
186.59 | |
RWOLDZZTBNYTMS-UHFFFAOYSA-N | |
97720 | |
C1=CC=C(C(=C1)C(C(=O)O)O)Cl |
C8H7ClO3 | |
MFCD00084962 | |
2-2-chlorophenyl-2-hydroxy-acetic acid, 2-2-chlorophenyl-2-hydroxyacetic acid, 2-chloro-mandelic acid, 2-chloromandelic acid, 2-chloromandelicacid, 2-chlorophenyl glycolic acid, 2-chlorophenyl hydroxy acetic acid, dl-2-chloro mandelic acid, dl-2-chloromandelic acid, pubchem15895 | |
2-(2-chlorophenyl)-2-hydroxyacetic acid |
Safety and Handling
GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.
GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Warning
EINECSNumber : 233-900-9