missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorolepidine, TCI America™

$59.58 - $203.20

Chemical Identifiers

CAS 634-47-9
Molecular Formula C10H8ClN
Molecular Weight (g/mol) 177.631
MDL Number MFCD00006742
InChI Key PFEIMKNQOIFKSW-UHFFFAOYSA-N
Synonym 2-chlorolepidine, 2-chloro-4-methyl-quinoline, quinoline, 2-chloro-4-methyl, lepidine, 2-chloro, pubchem5879, acmc-209nfp, 2-chloro-4methylquinoline, 4-methyl-2-chloroquinoline, 2-chlor-4-methyl-chinolin, 2-chloro-4-metyhylquinoline
PubChem CID 69449
IUPAC Name 2-chloro-4-methylquinoline
SMILES CC1=CC(=NC2=CC=CC=C12)Cl
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
C019125G View Documents TCI America
C019125G
25g Each for $203.20
 
C01915G View Documents TCI America
C01915G
5g Each for $59.58
 
Specifications

Chemical Identifiers

634-47-9
177.631
PFEIMKNQOIFKSW-UHFFFAOYSA-N
69449
CC1=CC(=NC2=CC=CC=C12)Cl
C10H8ClN
MFCD00006742
2-chlorolepidine, 2-chloro-4-methyl-quinoline, quinoline, 2-chloro-4-methyl, lepidine, 2-chloro, pubchem5879, acmc-209nfp, 2-chloro-4methylquinoline, 4-methyl-2-chloroquinoline, 2-chlor-4-methyl-chinolin, 2-chloro-4-metyhylquinoline
2-chloro-4-methylquinoline

Specifications

296°C
58°C
634-47-9
MFCD00006742
PFEIMKNQOIFKSW-UHFFFAOYSA-N
2-chloro-4-methylquinoline
69449
Crystal-Powder at 20°C
2-Chlorolepidine
White-Yellow
25g
C10H8ClN
2-chlorolepidine, 2-chloro-4-methyl-quinoline, quinoline, 2-chloro-4-methyl, lepidine, 2-chloro, pubchem5879, acmc-209nfp, 2-chloro-4methylquinoline, 4-methyl-2-chloroquinoline, 2-chlor-4-methyl-chinolin, 2-chloro-4-metyhylquinoline
CC1=CC(=NC2=CC=CC=C12)Cl
177.631
177.63
≥98.0% (GC)
Safety and Handling

Safety and Handling

EINECSNumber : (5)-0720

TSCA : No