Please login to your online account to display your discounted pricing

2-Chloroisonicotinaldehyde, 97%, Maybridge


Quantity 1g

Manufacturer: fisher scientific company  CC53604DA

 View more versions of this product

Catalog No. CC53604DA

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Formula Weight 141.56
Quantity 1g
Assay 97%
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H4ClNO
MDL Number MFCD06651557
CAS 101066-61-9
CAS Min % 96.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS P Statement Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful in contact with skin.
Chemical Name or Material 2-Chloroisonicotinaldehyde
Health Hazard 3 GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

Specifications

Chemical Name or Material 2-Chloroisonicotinaldehyde
CAS 101066-61-9
CAS Min % 96.5
CAS Max % 100.0
Assay 97%
Assay Percent Range 97%
Molecular Formula C6H4ClNO
Formula Weight 141.56
MDL Number MFCD06651557
Accessories

GHS Signal Word: Warning

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS P Statement Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful in contact with skin.