missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroisonicotinaldehyde, 97%, Maybridge

Chemical Identifiers

CAS 101066-61-9
CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Molecular Formula C6H4ClNO
Formula Weight 141.56
MDL Number MFCD06651557
Chemical Name or Material 2-Chloroisonicotinaldehyde
Assay 97%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
CC53604CB SDS Fisher Scientific Company
CC53604CB
250mg N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
CC53604DA SDS Fisher Scientific Company
CC53604DA
1g N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications

Description

2-Chloroisonicotinaldehyde, ≥97%
Description & Specifications

Chemical Identifiers

101066-61-9
100.0
C6H4ClNO
MFCD06651557
97%
96.5
97%
141.56
2-Chloroisonicotinaldehyde

Specifications

250mg
96.5
97%
141.56
2-Chloroisonicotinaldehyde
101066-61-9
100.0
C6H4ClNO
MFCD06651557
97%

Safety and Handling

GHS P Statement Harmful if inhaled. Harmful if swallowed. Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. Harmful in contact with skin.

GHS P Statement IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. IF ON SKIN: Wash with plenty of soap and water. Avoid breathing dust/fume/gas/mist/vapors/spray. IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell. IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. Wear protective gloves/protective clothing/eye protection/face protection.

GHS Signal Word: Warning