Learn More
2-Chloroethylphosphonic acid, 90%, Tech., Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 301630010
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloroethylphosphonic acid | |
16672-87-0 | |
100.0 | |
White to Beige | |
90% | |
C2H6ClO3P | |
1 g | |
ethephon, 2-chloroethyl phosphonic acid, ethrel, chlorethephon, bromeflor, camposan, 2-cepa, phosphonic acid, 2-chloroethyl, 2-chloroethanephosphonic acid, etheverse | |
UDPGUMQDCGORJQ-UHFFFAOYSA-N | |
2-chloroethylphosphonic acid | |
27982 | |
144.51 | |
Technical |
tech., 90% | |
88.0 | |
74.0°C to 75.0°C | |
Authentic | |
Glass bottle | |
MFCD00002142 | |
15, 3787 | |
Solubility in water: freely soluble. Other solubilities: slightly soluble in benzene,toluene,propylene glycol,practically insoluble in petroleum ether,practically insoluble in petroleum ether,freely soluble in ethanol,methanol,acetone,ethy | |
OP(O)(=O)CCCl | |
144.49 | |
CHEBI:52741 | |
90% | |
Powder |
Chemical Identifiers
16672-87-0 | |
144.49 | |
UDPGUMQDCGORJQ-UHFFFAOYSA-N | |
27982 | |
2-chloroethylphosphonic acid |
C2H6ClO3P | |
MFCD00002142 | |
ethephon, 2-chloroethyl phosphonic acid, ethrel, chlorethephon, bromeflor, camposan, 2-cepa, phosphonic acid, 2-chloroethyl, 2-chloroethanephosphonic acid, etheverse | |
CHEBI:52741 | |
OP(O)(=O)CCCl |
Safety and Handling
GHS H Statement
Harmful if swallowed.
Toxic in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Toxic to aquatic life with long lasting effects.
GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Immediately
GHS Signal Word: Danger
EINECSNumber : 240-718-3
RTECSNumber : SZ7100000
TSCA : TSCA
RUO – Research Use Only