missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-chloroethylephenyl sulfide, 98%, ACROS Organics™

Chemical Identifiers

CAS 5535-49-9
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Molecular Formula C8H9ClS
Formula Weight 172.68
Synonym 2-(Phenylthio)ethylechloride
Chemical Name or Material 2-chloroethylephenyl sulfide
Assay ≥98.5%
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
Catalog Number Mfr. No. Packaging Quantity Price Quantity  
AC302210050 SDS Thermo Fisher Scientific
AC302210050
Glass bottle 5mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

5535-49-9
100.0
C8H9ClS
2-(Phenylthio)ethylechloride
≥98.5%
97.5
98%
172.68
2-chloroethylephenyl sulfide

Specifications

Liquid
5mL
1.17g/mL
Authentic
97.5
98%
2-(Phenylthio)ethylechloride
2-chloroethylephenyl sulfide
172.68
Clear colorless
100.0
1.5820 to 1.5840 (20°C, 589nm)
5535-49-9
C8H9ClS
Glass bottle
≥98.5%