Please login to your online account to display your discounted pricing

2-chloroethylephenyl sulfide, 98%, ACROS Organics™


Quantity 5mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC302210050

 View more versions of this product

Catalog No. AC302210050

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥98.5%
Physical Form Liquid
Formula Weight 172.68
Quantity 5mL
Color Clear colorless
Density 1.17g/mL
CAS Max % 100.0
Infrared Spectrum Authentic
Refractive Index 1.5820 to 1.5840 (20°C, 589nm)
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material 2-chloroethylephenyl sulfide
CAS Min % 97.5
CAS 5535-49-9
Assay Percent Range 98%
Molecular Formula C8H9ClS
Synonym 2-(Phenylthio)ethylechloride
Packaging Glass bottle
Health Hazard 1 GHS Signal Word: Danger

Specifications

Chemical Name or Material 2-chloroethylephenyl sulfide
CAS 5535-49-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥98.5%
Assay Percent Range 98%
Molecular Formula C8H9ClS
Formula Weight 172.68
Synonym 2-(Phenylthio)ethylechloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Immediately call a POISON CENTER or doctor/physician.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF ON SKIN: Wash with plenty of soap and water.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Harmful in contact with skin.
Harmful if inhaled.
Causes severe skin burns and eye damage.

SDS & Safety Documents