Please login to your online account to display your discounted pricing

2-chloroethyleisothiocyanate, 98%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-chloroethyleisothiocyanate
CAS 6099-88-3
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H4ClNS
Linear Formula ClCHCHNCS
Formula Weight 121.59
MDL Number MFCD00041125
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC309430010

  SDS
thermo fisher scientific
AC309430010
Glass bottle 1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Physical Form Liquid
Assay ≥97.5%
Formula Weight 121.59
Quantity 1g
Color Clear yellow
CAS Max % 100.0
Packaging Glass bottle
Refractive Index 1.5550 to 1.5570 (20°C, 589nm)
Linear Formula ClCHCHNCS
Chemical Name or Material 2-chloroethyleisothiocyanate
Assay Percent Range 98%
Molecular Formula C3H4ClNS
MDL Number MFCD00041125
CAS 6099-88-3
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Warning
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful in contact with skin.
Causes serious eye irritation.
Flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Infrared Spectrum Authentic

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your 9-digit account number, or become a registered user on fishersci.com by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service at 1-800-766-7000.