Please login to your online account to display your discounted pricing

2-Chloroethyl Vinyl Ether (For EPA 600, 8000/Appendix IX Methods)

$58.23 - $58.23

Chemical Identifiers

Chemical Name or Material 2-Chloroethyl Vinyl Ether
CAS 110-75-8
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

US-HC-070-1

 
ultra scientific
HC0701
Ampule 1x1mL Each for $58.23
Description & Specifications

Specifications

Quantity 1x1mL
Alpha Vector Chloroethyl Vinyl Ether
Methods For EPA 600, 8000/ Appendix IX Methods
Chemical Name or Material 2-Chloroethyl Vinyl Ether
Packaging Ampule
CAS 110-75-8