Learn More
2-Chloroethyl vinyl ether, ca. 95%, Thermo Scientific™
Supplier: Thermo Scientific Chemicals 149431000
Description
This Thermo Scientific brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Specifications
2-Chloroethyl vinyl ether | |
110-75-8 | |
100.0 | |
109°C | |
Authentic | |
Glass bottle | |
1.4370 to 1.439 | |
MFCD00000973 | |
01, II, 473 | |
15, 2140 | |
Solubility in water: slightly soluble. Other solubilities: hydrolysis in alkaline waters | |
C=COCCCl | |
106.55 | |
106.55 |
ca. 95% | |
94.0 | |
1.0480g/mL | |
16°C | |
94% min. (GC) | |
C4H7ClO | |
ClCH2CH2OCH=CH2 | |
100g | |
1.048 | |
2-chloroethyl vinyl ether, 2-chloroethoxy ethene, ethene, 2-chloroethoxy, 2-vinyloxyethyl chloride, vinyl 2-chloroethyl ether, ether, 2-chloroethyl vinyl, rcra waste number u042, beta-chloroethyl vinyl ether, 2-chlorethyl vinyl ether | |
DNJRKFKAFWSXSE-UHFFFAOYSA-N | |
1-chloro-2-ethenoxyethane | |
8074 | |
ca. 95% |
Safety and Handling
GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Flammable liquid and vapor.
Highly flammable liquid and vapor.
GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.
WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
GHS Signal Word: Danger
EINECSNumber : 203-799-6
RTECSNumber : KN6300000
TSCA : TSCA
RUO â Research Use Only