missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chloroethyl vinyl ether, ca. 95%, ACROS Organics™

Chemical Identifiers

CAS 110-75-8
Molecular Formula C4H7ClO
Molecular Weight (g/mol) 106.55
MDL Number MFCD00000973
InChI Key DNJRKFKAFWSXSE-UHFFFAOYSA-N
Synonym 2-chlorethyl vinyl ether, 2-chloroethoxy ethene, 2-chloroethyl vinyl ether, 2-vinyloxyethyl chloride, beta-chloroethyl vinyl ether, ethene, 2-chloroethoxy, ether, 2-chloroethyl vinyl, rcra waste number u042, vinyl 2-chloroethyl ether
PubChem CID 8074
IUPAC Name 1-chloro-2-ethenoxyethane
SMILES C=COCCCl
View More Specs

Products 1
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
Catalog Number Mfr. No. Quantity Packaging Price Quantity  
AC149431000
SDS View Documents
Thermo Fisher Scientific
149431000
100g Glass bottle N/A
N/A
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description

Description

Specifications

Chemical Identifiers

110-75-8
106.55
DNJRKFKAFWSXSE-UHFFFAOYSA-N
8074
C=COCCCl
C4H7ClO
MFCD00000973
2-chlorethyl vinyl ether, 2-chloroethoxy ethene, 2-chloroethyl vinyl ether, 2-vinyloxyethyl chloride, beta-chloroethyl vinyl ether, ethene, 2-chloroethoxy, ether, 2-chloroethyl vinyl, rcra waste number u042, vinyl 2-chloroethyl ether
1-chloro-2-ethenoxyethane

Specifications

1.0480g/mL
Authentic
1.4370 to 1.439
109°C
-70°C
110-75-8
100.0
C4H7ClO
MFCD00000973
15, 2140
Solubility in water: slightly soluble. Other solubilities: hydrolysis in alkaline waters
C=COCCCl
106.55
106.55
ca. 95%
16°C
Glass bottle
1.048
Colorless to Yellow
100g
94.0
94% min. (GC)
ClCH2CH2OCH=CH2
01, II, 473
2-chlorethyl vinyl ether, 2-chloroethoxy ethene, 2-chloroethyl vinyl ether, 2-vinyloxyethyl chloride, beta-chloroethyl vinyl ether, ethene, 2-chloroethoxy, ether, 2-chloroethyl vinyl, rcra waste number u042, vinyl 2-chloroethyl ether
DNJRKFKAFWSXSE-UHFFFAOYSA-N
1-chloro-2-ethenoxyethane
8074
Liquid
2-Chloroethyl vinyl ether, ca. 95%
Safety and Handling

Safety and Handling

GHS H Statement:
May cause respiratory irritation.
Harmful if inhaled.
Causes serious eye damage.
Causes skin irritation.
Flammable liquid and vapor.
Highly flammable liquid and vapor.

GHS P Statement:
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 203-799-6

RTECSNumber : KN6300000

TSCA : TSCA

SDS