missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Chloroethyl)triphenylphosphonium bromide, 98+%

$84.68 - $303.80

Chemical Identifiers

CAS 31238-20-7
Molecular Formula C20H19BrClP
Molecular Weight (g/mol) 405.7
MDL Number MFCD00051877
InChI Key HETXOZDAMZALJQ-UHFFFAOYSA-M
Synonym 2-chloroethyl triphenylphosphonium bromide, 2-chloroethyltriphenylphosphonium bromide, 2-chloroethyl triphenylphosphanium bromide, acmc-1cp77, 2-chloroethyl triphenyl phosphanium bromide, 2-chloroethyl triphenyl phosphonium bromide, phosphonium, 2-chloroethyl triphenyl-, bromide 1:1
PubChem CID 3084530
IUPAC Name 2-chloroethyl(triphenyl)phosphanium;bromide
SMILES C1=CC=C(C=C1)[P+](CCCl)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AAL0053809 View Documents Alfa Aesar™
L0053809
10g Each for $84.68
 
AAL0053818 View Documents Alfa Aesar™
L0053818
50g Each for $303.80
 
Specifications

Chemical Identifiers

31238-20-7
405.7
HETXOZDAMZALJQ-UHFFFAOYSA-M
3084530
C1=CC=C(C=C1)[P+](CCCl)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
C20H19BrClP
MFCD00051877
2-chloroethyl triphenylphosphonium bromide, 2-chloroethyltriphenylphosphonium bromide, 2-chloroethyl triphenylphosphanium bromide, acmc-1cp77, 2-chloroethyl triphenyl phosphanium bromide, 2-chloroethyl triphenyl phosphonium bromide, phosphonium, 2-chloroethyl triphenyl-, bromide 1:1
2-chloroethyl(triphenyl)phosphanium;bromide

Specifications

169°C to 172°C
31238-20-7
MFCD00051877
HETXOZDAMZALJQ-UHFFFAOYSA-M
2-chloroethyl(triphenyl)phosphanium;bromide
3084530
≥98%
10g
C20H19BrClP
2-chloroethyl triphenylphosphonium bromide, 2-chloroethyltriphenylphosphonium bromide, 2-chloroethyl triphenylphosphanium bromide, acmc-1cp77, 2-chloroethyl triphenyl phosphanium bromide, 2-chloroethyl triphenyl phosphonium bromide, phosphonium, 2-chloroethyl triphenyl-, bromide 1:1
C1=CC=C(C=C1)[P+](CCCl)(C2=CC=CC=C2)C3=CC=CC=C3.[Br-]
405.7
405.7
(2-Chloroethyl)triphenylphosphonium bromide
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning