missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ (2-Chloroethyl)trimethylammonium chloride, 98% (dry wt.), may cont. up to 5% water

Manufacturer:  Alfa Aesar™ A1563006

 View more versions of this product

Catalog No. AAA1563006



Specifications

Specifications

(2-Chloroethyl)trimethylammonium chloride
~241°C (decomposition)
999-81-5
C5H13Cl2N
MFCD00011869
3563994
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
C[N+](C)(C)CCCl.[Cl-]
158.066
158.07
Amine-like
5g
(dry wt.), may cont. up to 5% water
ClCH2CH2N(CH3)3Cl
UN2811
14,2104
UHZZMRAGKVHANO-UHFFFAOYSA-M
2-chloroethyl(trimethyl)azanium;chloride
13836
98%

Chemical Identifiers

999-81-5
158.066
UHZZMRAGKVHANO-UHFFFAOYSA-M
13836
C[N+](C)(C)CCCl.[Cl-]
C5H13Cl2N
MFCD00011869
chlormequat chloride, 2-chloro-n,n,n-trimethylethanaminium chloride, chlormequat, chlorocholine chloride, antywylegacz, cycocel, 2-chloroethyl trimethylammonium chloride, cyclocel, stabilan, cycogan
2-chloroethyl(trimethyl)azanium;chloride
Safety and Handling

Safety and Handling

GHS H Statement
H311-H302
Toxic in contact with skin.
Harmful if swallowed.

GHS P Statement
P280-P361-P301+P312-P312-P405-P501a
Wear protective gloves/protective clothing/eye protection/face protection.
Remove/Take off immediately all contaminated clothing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : BP5250000

TSCA : TSCA

Documents