Learn More
2-Chloroethyl phenyl sulfone, 98%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals B2169014
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloroethyl phenyl sulfone | |
52°C to 55°C | |
C8H9ClO2S | |
25 g | |
1104341 | |
NUJGORANFDSMOL-UHFFFAOYSA-N | |
2-chloroethylsulfonylbenzene | |
13646 | |
98% |
938-09-0 | |
168°C to 174°C (7 mmHg) | |
MFCD00025047 | |
UN1759 | |
2-chloroethyl phenyl sulfone, 2-chloroethanesulfonyl benzene, 2-chloroethyl sulfonyl benzene, 2-chloroethylphenylsulfone, 2-chloroethylsulfonyl benzene, sulfone, 2-chloroethyl phenyl, 2-chloroethyl sulphonyl benzene, benzene, 2-chloroethyl sulfonyl, 2-chloroethylphenyl sulfone | |
C1=CC=C(C=C1)S(=O)(=O)CCCl | |
204.668 | |
204.68 |
Chemical Identifiers
938-09-0 | |
204.668 | |
NUJGORANFDSMOL-UHFFFAOYSA-N | |
13646 | |
C1=CC=C(C=C1)S(=O)(=O)CCCl |
C8H9ClO2S | |
MFCD00025047 | |
2-chloroethyl phenyl sulfone, 2-chloroethanesulfonyl benzene, 2-chloroethyl sulfonyl benzene, 2-chloroethylphenylsulfone, 2-chloroethylsulfonyl benzene, sulfone, 2-chloroethyl phenyl, 2-chloroethyl sulphonyl benzene, benzene, 2-chloroethyl sulfonyl, 2-chloroethylphenyl sulfone | |
2-chloroethylsulfonylbenzene |
Safety and Handling
GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, N.O.S.
EINECSNumber : 213-337-5
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only