Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroethyl p-tolyl sulfone, 98%

$67.58 - $218.58

Chemical Identifiers

Chemical Name or Material 2-Chloroethyl p-tolyl sulfone
CAS 22381-53-9
Molecular Formula C9H11ClO2S
Formula Weight 218.7
MDL Number MFCD00051607
Beilstein 1107627
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL0525603

 
alfa aesar™
L0525603
1g Each for $67.58

AAL0525606

 
alfa aesar™
L0525606
5g Each for $218.58
Description & Specifications

Specifications

Formula Weight 218.7
Chemical Name or Material 2-Chloroethyl p-tolyl sulfone
MDL Number MFCD00051607
CAS 22381-53-9
Assay 98%
Molecular Formula C9H11ClO2S
Beilstein 1107627
Melting Point 77° to 81°C
Quantity 1g