Please login to your online account to display your discounted pricing

2-Chloroethyl-p-toluenesulfonate 97%, ACROS Organics™

Chemical Identifiers

Chemical Name or Material 2-Chloroethyl-p-toluenesulfonate
Name Note 97%
CAS 80-41-1
CAS Min % 96.0
CAS Max % 100.0
Assay ≥96.0%
Assay Percent Range 97%
Molecular Formula C9H11ClO3S
Linear Formula CH3C6H4SO3CH2CH2Cl
Formula Weight 234.7
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC148381000

  SDS
thermo fisher scientific
148381000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AC148380250

  SDS
thermo fisher scientific
148380250
Glass bottle 25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥96.0%
Physical Form Liquid
Formula Weight 234.7
Quantity 100g
Color Clear colorless to yellow
EINECS Number 201-277-2
TSCA TSCA
RTECS Number XT6475000
Beilstein 11, II, 45
CAS Min % 96.0
CAS Max % 100.0
Chemical Name or Material 2-Chloroethyl-p-toluenesulfonate
Refractive Index 1.5280 to 1.5300 (20°C, 589nm)
MDL Number MFCD00000970
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Packaging Glass bottle
Name Note 97%
Assay Percent Range 97%
Molecular Formula C9H11ClO3S
Linear Formula CH3C6H4SO3CH2CH2Cl
CAS 80-41-1
Infrared Spectrum Authentic
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Safety and Handling

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.