missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
2-Chloroethyl p-toluenesulfonate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals A1186314
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
2-Chloroethyl p-toluenesulfonate | |
1.294 | |
>110°C (230°F) | |
1.529 | |
25 g | |
2-chloroethyl p-toluenesulfonate, 2-chloroethyl tosylate, 2-chloroethyl p-tosylate, 2-tosyloxyethyl chloride, 2-chloroethanol p-toluenesulfonate, p-toluenesulfonic acid, 2-chloroethyl ester, ethanol, 2-chloro-, 4-methylbenzenesulfonate, 2-p-toluenesulfonyloxy ethyl chloride, 2-chloroethyl-p-toluenesulfonate, p-toluenesulfonic acid-2-chloroethyl ester | |
CC1=CC=C(C=C1)S(=O)(=O)OCCCl | |
234.694 | |
234.7 |
80-41-1 | |
158°C to 159°C (0.5 mmHg) | |
C9H11ClO3S | |
MFCD00000970 | |
1968816 | |
ZXNMIUJDTOMBPV-UHFFFAOYSA-N | |
2-chloroethyl 4-methylbenzenesulfonate | |
6639 | |
97% |
Chemical Identifiers
80-41-1 | |
234.694 | |
ZXNMIUJDTOMBPV-UHFFFAOYSA-N | |
6639 | |
CC1=CC=C(C=C1)S(=O)(=O)OCCCl |
C9H11ClO3S | |
MFCD00000970 | |
2-chloroethyl p-toluenesulfonate, 2-chloroethyl tosylate, 2-chloroethyl p-tosylate, 2-tosyloxyethyl chloride, 2-chloroethanol p-toluenesulfonate, p-toluenesulfonic acid, 2-chloroethyl ester, ethanol, 2-chloro-, 4-methylbenzenesulfonate, 2-p-toluenesulfonyloxy ethyl chloride, 2-chloroethyl-p-toluenesulfonate, p-toluenesulfonic acid-2-chloroethyl ester | |
2-chloroethyl 4-methylbenzenesulfonate |
Safety and Handling
GHS H Statement
H302
Harmful if swallowed.
P264b-P270-P301+P312-P330-P501c
H302
EINECSNumber : 201-277-2
RTECSNumber : XT6475000
TSCA : Yes
Recommended Storage : Keep cold
RUO – Research Use Only