missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroethyl p-toluenesulfonate, 97%

Manufacturer:  Alfa Aesar™ A1186322

 View more versions of this product

Catalog No. AAA1186322


 

Specifications

Specifications

2-Chloroethyl p-toluenesulfonate
158°C to 159°C (0.5mmHg)
80-41-1
C9H11ClO3S
1968816
ZXNMIUJDTOMBPV-UHFFFAOYSA-N
2-chloroethyl 4-methylbenzenesulfonate
6639
>110°C (230°F)
100g
97%
MFCD00000970
p-Toluenesulfonic acid 2-chloroethyl ester
CC1=CC=C(C=C1)S(=O)(=O)OCCCl
234.694
234.7

Chemical Identifiers

80-41-1
234.694
ZXNMIUJDTOMBPV-UHFFFAOYSA-N
6639
C9H11ClO3S
MFCD00000970
p-Toluenesulfonic acid 2-chloroethyl ester
Safety and Handling

Safety and Handling

GHS H Statement
H302
Harmful if swallowed.

GHS P Statement
P280f
Wear protective clothing.

Warning

RTECSNumber : XT6475000

TSCA : TSCA

Recommended Storage : 2°C to 8°C

SDS Documents