Please login to your online account to display your discounted pricing

2-Chloroethyl methyl ether, 98%, ACROS Organics™


Quantity 500mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221775000

 View more versions of this product

Catalog No. AC221775000

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.5%
Physical Form Liquid
Formula Weight 94.54
Quantity 500mL
Color Clear colorless to yellow
EINECS Number 210-999-7
TSCA TSCA
CAS Max % 100.0
Chemical Name or Material 2-Chloroethyl methyl ether
Name Note 98%
Assay Percent Range 98%
Molecular Formula C3H7ClO
Linear Formula ClCH2CH2OCH3
MDL Number MFCD00000972
CAS 627-42-9
CAS Min % 97.5
Health Hazard 1 GHS Signal Word: Danger
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4080 to 1.4100 (20°C, 589nm)
Water 0.2% max. (K.F.)
Synonym 2-Methoxyethyl chloride
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.
Health Hazard 3 GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Chloroethyl methyl ether
Name Note 98%
CAS 627-42-9
CAS Min % 97.5
CAS Max % 100.0
Assay ≥97.5%
Assay Percent Range 98%
Molecular Formula C3H7ClO
Linear Formula ClCH2CH2OCH3
Formula Weight 94.54
MDL Number MFCD00000972
Synonym 2-Methoxyethyl chloride
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Ground/Bond container and receiving equipment.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Highly flammable liquid and vapor.

SDS & Safety Documents