Learn More
2-Chloroethyl methacrylate, 97%, stab. with 400-600ppm hydroquinone, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0978309
Description
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).
Solubility
Immiscible with water.
Notes
Incompatible with oxidizing agents.
Specifications
2-Chloroethyl methacrylate | |
1888-94-4 | |
69°C (156°F) | |
1.451 | |
10 g | |
956599 | |
Immiscible with water. | |
CC(=C)C(=O)OCCCl | |
148.59 | |
148.59 |
stabilized with 500ppm hydroquinone | |
170°C | |
C6H9ClO2 | |
MFCD00045291 | |
UN2810 | |
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate | |
GPOGMJLHWQHEGF-UHFFFAOYSA-N | |
2-chloroethyl 2-methylprop-2-enoate | |
15905 | |
97% |
Chemical Identifiers
1888-94-4 | |
148.59 | |
GPOGMJLHWQHEGF-UHFFFAOYSA-N | |
15905 | |
CC(=C)C(=O)OCCCl |
C6H9ClO2 | |
MFCD00045291 | |
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate | |
2-chloroethyl 2-methylprop-2-enoate |
Safety and Handling
GHS H Statement
H301-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P235-P260-P264b-P270-P271-P280-P284-P301+P310-P302+P352-P304+P340-P305+P351+P338-P310-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H315-H319-H330-H335
DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.
EINECSNumber : 217-566-1
RTECSNumber : OZ3855000
TSCA : Yes
Recommended Storage : Ambient temperatures
RUO – Research Use Only