missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroethyl methacrylate, 97%, stab. with 500ppm hydroquinone

Manufacturer:  Alfa Aesar™ L0978318

 View more versions of this product

Catalog No. AAL0978318



Description

Description

Applications
2-Chloroethyl methacrylate is used as a monomer to prepare poly(2-chloroethyl methacrylate).

Solubility
Immiscible with water.

Notes
Incompatible with oxidizing agents.
Specifications

Specifications

2-Chloroethyl methacrylate
170°C
stabilized with 500ppm hydroquinone
C6H9ClO2
UN2810
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate
CC(=C)C(=O)OCCCl
148.586
148.59
69°C (156°F)
50g
1888-94-4
MFCD00045291
956599
GPOGMJLHWQHEGF-UHFFFAOYSA-N
2-chloroethyl 2-methylprop-2-enoate
15905
97%

Chemical Identifiers

1888-94-4
148.586
GPOGMJLHWQHEGF-UHFFFAOYSA-N
15905
CC(=C)C(=O)OCCCl
C6H9ClO2
MFCD00045291
2-chloroethyl methacrylate, chloroethyl methacrylate, methacrylic acid, 2-chloroethyl ester, 2-propenoic acid, 2-methyl-, 2-chloroethyl ester, .beta.-chloroethyl methacrylate, methacrylic acid beta-chloroethyl ester, methacrylic acid .beta.-chloroethyl ester, acmc-20anxx, chloroethylmethacrylate, 2-chloroethylmethacrylate
2-chloroethyl 2-methylprop-2-enoate
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H227-H315-H319-H335
Toxic if swallowed.
Fatal if inhaled.
Combustible liquid.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P210-P301+P310-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: TOXIC, LIQUIDS, ORGANIC, N.O.S.

RTECSNumber : OZ3855000

TSCA : TSCA

Documents