Please login to your online account to display your discounted pricing

2-Chloroethyl ethyl sulfide, 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  221970250

 View more versions of this product

Catalog No. AC221970250

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥94% (GC)
Physical Form Liquid
Formula Weight 124.63
Quantity 25g
Color Clear colorless to light yellow
Density 1.07g/mL
Chemical Name or Material 2-Chloroethyl ethyl sulfide
Packaging Glass bottle
Refractive Index 1.4875 to 1.4895 (20°C, 589nm)
Assay Percent Range 95%
Molecular Formula C4H9ClS
Linear Formula ClCH2CH2SC2H5
CAS 693-07-2
Infrared Spectrum Authentic

Specifications

Chemical Name or Material 2-Chloroethyl ethyl sulfide
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C4H9ClS
Linear Formula ClCH2CH2SC2H5
Formula Weight 124.63