Please login to your online account to display your discounted pricing

2-Chloroethyl ethyl sulfide, 95%, ACROS Organics™

$39.11 - $133.77

Chemical Identifiers

Chemical Name or Material 2-Chloroethyl ethyl sulfide
Assay ≥94% (GC)
Assay Percent Range 95%
Molecular Formula C4H9ClS
Linear Formula ClCH2CH2SC2H5
Formula Weight 124.63
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

AC221970050

  SDS
thermo fisher scientific
221970050
Glass bottle 5g Each for $39.11

AC221970250

  SDS
thermo fisher scientific
221970250
Glass bottle 25g Each for $133.77

AC221971000

  SDS
thermo fisher scientific
AC221971000
Glass bottle 100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Assay ≥94% (GC)
Physical Form Liquid
Formula Weight 124.63
Quantity 5g
Color Clear colorless to light yellow
Density 1.07g/mL
Refractive Index 1.4875 to 1.4895 (20°C, 589nm)
CAS 693-07-2
Linear Formula ClCH2CH2SC2H5
Chemical Name or Material 2-Chloroethyl ethyl sulfide
Packaging Glass bottle
Assay Percent Range 95%
Molecular Formula C4H9ClS
Infrared Spectrum Authentic