missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-(Chloroethyl)benzene, 99%, Thermo Scientific Chemicals

Supplier:  Thermo Scientific Chemicals L0489122

 View more versions of this product

Catalog No. AAL0489122



Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Specifications

2-(Chloroethyl)benzene
-60°C
82°C to 84°C (16 mmHg)
C8H9Cl
MFCD00000977
385969
MNNZINNZIQVULG-UHFFFAOYSA-N
2-chloroethylbenzene
231496
99%
622-24-2
1.07
66°C (150°F)
1.53
100 g
2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
ClCCC1=CC=CC=C1
140.61
140.61

Chemical Identifiers

622-24-2
140.61
MNNZINNZIQVULG-UHFFFAOYSA-N
231496
ClCCC1=CC=CC=C1
C8H9Cl
MFCD00000977
2-chloroethyl benzene, phenethyl chloride, 2-phenylethyl chloride, benzene, 2-chloroethyl, 1-chloro-2-phenylethane, phenethylchloride, ethylchlorobenzene, 2-phenyl-1-chloroethane, 2-chloroethyl-benzene, beta-phenethyl chloride
2-chloroethylbenzene
Safety and Handling

Safety and Handling

GHS H Statement
H227
Combustible liquid.

P210-P235-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c

H227-H315-H319-H335

EINECSNumber : 210-725-6

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only