Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroethyl 4-fluorophenyl sulfone, 98%

$62.34 - $232.43

Chemical Identifiers

Chemical Name or Material 2-Chloroethyl 4-fluorophenyl sulfone
CAS 33330-46-0
Molecular Formula C8H8ClFO2S
Formula Weight 222.67
MDL Number MFCD00203483
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1693003

  SDS
alfa aesar™
A1693003
1g Each for $62.34

AAA1693006

  SDS
alfa aesar™
A1693006
5g Each for $232.43
Description & Specifications

Specifications

Formula Weight 222.67
Molecular Formula C8H8ClFO2S
MDL Number MFCD00203483
CAS 33330-46-0
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Melting Point 67° to 70°C
Quantity 1g
Chemical Name or Material 2-Chloroethyl 4-fluorophenyl sulfone
Assay 98%
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.