missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chloroethanesulfonyl chloride, 98%

Manufacturer:  Alfa Aesar™ A1537730

 View more versions of this product

Catalog No. AAA1537730



Description

Description

Applications
2-Chloroethanesulfonyl chloride was used in one-pot sulfonylation/intramolecular thia-Michael protocol for the synthesis of 1,5,2-dithiazepine 1,1-dioxides. It was also used in synthesis of vinyl sulfonamides with a furan, carbocyclic, semi cyclic or acyclic 1,3-diene moiety.

Solubility
Reacts with water.

Notes
Moisture sensitive. Store under inert gas. Store away from moisture, oxidizing agents. Store in refrigerator.
Specifications

Specifications

2-Chloroethanesulfonyl chloride
203°C
1622-32-8
ClCH2CH2SO2Cl
UN3390
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
C(CCl)S(=O)(=O)Cl
163.012
163.02
>110°C (230°F)
250g
C2H4Cl2O2S
MFCD00007461
1751202
VHCSBTPOPKFYIU-UHFFFAOYSA-N
2-chloroethanesulfonyl chloride
15385
98%

Chemical Identifiers

1622-32-8
163.012
VHCSBTPOPKFYIU-UHFFFAOYSA-N
15385
C(CCl)S(=O)(=O)Cl
C2H4Cl2O2S
MFCD00007461
2-chloroethyl sulphonyl chloride, 2-chloroethylsulfonyl chloride, ethanesulfonyl chloride, 2-chloro, 2-chloroethane sulfochloride, 2-chloro-ethanesulfonyl chloride, 2-chloroethanesulfonylchloride, beta-chloroethanesulfonyl chloride, 2-chloroethane-1-sulfonyl chloride, ethanesulfonylchloride, 2-chloro, 2-chloro-1-ethanesulfonyl chloride
2-chloroethanesulfonyl chloride
Safety and Handling

Safety and Handling

GHS H Statement
H301-H330-H314-H318
Toxic if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P301+P310-P303+P361+P353-P304+P340-P305+P351+P338-P320-P330-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Specific treatment is urgent (see label).
Rinse mouth.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: I; Proper Shipping Name: TOXIC BY INHALATION LIQUID, CORROSIVE, N.O.S.

RTECSNumber : KI8060000

TSCA : TSCA

Recommended Storage : 2° to 8°C

Documents