Please login to your online account to display your discounted pricing

2-Chloroethanesulfonyl Chloride 95%, ACROS Organics™


Quantity 25g
Packaging Glass bottle

Manufacturer: thermo fisher scientific  404610250

 View more versions of this product

Catalog No. AC404610250

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥94%
Formula Weight 163.02
Physical Form Liquid
Quantity 25g
Color Clear slightly yellow to brown
TSCA TSCA
EINECS Number 216-594-1
RTECS Number KI8060000
CAS Min % 94.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
CAS 1622-32-8
Health Hazard 1 GHS Signal Word: Danger
Health Hazard 2 GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.
Molecular Formula C2H4Cl2O2S
Name Note 95%
Chemical Name or Material 2-Chloroethanesulfonyl chloride
Linear Formula ClCH2CH2SO2Cl
Assay Percent Range 95%
MDL Number MFCD00007461
Refractive Index 1.4910 to 1.4940 (20°C, 589nm)
Health Hazard 3 GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

Specifications

Chemical Name or Material 2-Chloroethanesulfonyl chloride
Name Note 95%
CAS 1622-32-8
CAS Min % 94.0
CAS Max % 100.0
Assay ≥94%
Assay Percent Range 95%
Molecular Formula C2H4Cl2O2S
Linear Formula ClCH2CH2SO2Cl
Formula Weight 163.02
MDL Number MFCD00007461
Accessories

GHS Signal Word: Danger

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Store in a dry place. Store in a closed container.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Harmful if swallowed.
Fatal if inhaled.
Causes severe skin burns and eye damage.
Contact with water liberates toxic gas.

SDS & Safety Documents