missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one, 97%, Alfa Aesar™

Manufacturer:  Thermo Fisher Scientific H54266MD

 View more versions of this product

Catalog No. AAH54266MD



Specifications

Specifications

2-Chlorodibenzo[b,f]-1,4-oxazepin-11(10H)-one
3158-91-6
MFCD06658244
ZAGINEPNYIZLLO-UHFFFAOYSA-N
8-chloro-5H-benzo[b][1,4]benzoxazepin-6-one
13000266
97%
250mg
C13H8ClNO2
13-chloro-2-oxa-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3 8 ,4,6,11,13-hexaen-10-one
C1=CC=C2C(=C1)NC(=O)C3=C(O2)C=CC(=C3)Cl
245.662
245.66

Chemical Identifiers

3158-91-6
245.662
ZAGINEPNYIZLLO-UHFFFAOYSA-N
13000266
C1=CC=C2C(=C1)NC(=O)C3=C(O2)C=CC(=C3)Cl
C13H8ClNO2
MFCD06658244
13-chloro-2-oxa-9-azatricyclo 9.4.0.0 3 ,? pentadeca-1 15 ,3 8 ,4,6,11,13-hexaen-10-one
8-chloro-5H-benzo[b][1,4]benzoxazepin-6-one
Safety and Handling

Safety and Handling

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning

EINECSNumber : 221-602-1

TSCA : No

Recommended Storage : Ambient temperatures

Documents