Please login to your online account to display your discounted pricing

Alfa Aesar™ 2-Chlorocyclopentanone, 98%, stab. with potassium carbonate

$85.43 - $85.43

Chemical Identifiers

Chemical Name or Material 2-Chlorocyclopentanone
CAS 694-28-0
Name Note stabilized with potassium carbonate
Molecular Formula C5H7ClO
Formula Weight 118.56
MDL Number MFCD00001410
Beilstein 1098899
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2257306

 
alfa aesar™
B2257306
5g Each for $85.43
Description & Specifications

Specifications

Formula Weight 118.56
Chemical Name or Material 2-Chlorocyclopentanone
TSCA TSCA
Name Note stabilized with potassium carbonate
Assay 98%
Molecular Formula C5H7ClO
MDL Number MFCD00001410
CAS 694-28-0
Beilstein 1098899
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
RTECS Number GY4900000
Flash Point 77°C (171°F)
Boiling Point 72° to 74°C (12mmHg)
Quantity 5g

Applications
2-Chlorocyclopentanone is used as pharmaceutical intermediate.

Solubility
Soluble in water.

Notes
Stable under recommended storage conditions. Incompatible with oxidizing agents.

Safety and Handling

Warning

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

−30° to −10°C